Gene Transcripts
- Gene Symbol
- TRIM36
- Entrez Gene ID
- 55521
- Gene Name
- tripartite motif containing 36
- Gene Aliases
- HAPRIN, RBCC728, RNF98
- Chromosome Location
- Chr. 5: 115124762 - 115180546 on Build GRCh38
- UniGene ID
- Hs.519514
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
GenBank mRNA |
AK125426.1 |
- |
1 |
1483 |
|
View Full Product Details