Gene Transcripts
- Gene Symbol
- SLC39A4
- Entrez Gene ID
- 55630
- Gene Name
- solute carrier family 39 member 4
- Gene Aliases
- AEZ, AWMS2, ZIP4
- Chromosome Location
- Chr. 8: 144412414 - 144416895 on Build GRCh38
- UniGene ID
- Hs.521934
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
GenBank mRNA |
AK056900.1 |
- |
3 |
2746 |
|
Non Gene Transcripts
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
Transcript Location |
GenBankmRNA |
AK055137.1 |
- |
1 |
1752 |
|
Chr. X - 48554573 - 48556746 [+] |
View Full Product Details