Gene Transcripts
- Gene Symbol
- ESRP2
- Entrez Gene ID
- 80004
- Gene Name
- epithelial splicing regulatory protein 2
- Gene Aliases
- RBM35B
- Chromosome Location
- Chr. 16: 68228547 - 68236233 on Build GRCh38
- UniGene ID
- Hs.436585
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
GenBank mRNA |
BU934976.1 |
- |
1 |
281 |
|
- Gene Symbol
- NFATC3
- Entrez Gene ID
- 4775
- Gene Name
- nuclear factor of activated T-cells 3
- Gene Aliases
- NFAT4, NFATX
- Chromosome Location
- Chr. 16: 68085366 - 68229259 on Build GRCh38
- UniGene ID
- Hs.436585
- Species
- Homo sapiens
Non Gene Transcripts
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
Transcript Location |
GenBankmRNA |
BG283936.1 |
- |
1 |
560 |
|
Chr. 16 - 68228137 - 68228831 [+] |
GenBankmRNA |
CR607695.1 |
- |
1 |
1473 |
|
Chr. 16 - 68227226 - 68229145 [+] |
GenBankmRNA |
BX327070.1 |
- |
1 |
567 |
|
Chr. 16 - 68228132 - 68229030 [+] |
GenBankmRNA |
BY995433.1 |
- |
1 |
683 |
|
Chr. 16 - 68228033 - 68228750 [+] |
GenBankmRNA |
CD103418.1 |
- |
|
497 |
|
Chr. - - [-] |
View Full Product Details