Gene Transcripts

Gene Symbol
Six3os1
Entrez Gene ID
100043902
Gene Name
SIX homeobox 3, opposite strand 1
Gene Aliases
D17Mgi26, E130112H22Rik, Rncr1, Six3os
Chromosome Location
Chr. 17: 85601932 - 85618396 on Build GRCm38
UniGene ID
Mm.150838
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NR_038082.1 4 2394
NR_038083.1 4 1478
GenBank mRNA AK053722.1 - 4 1480
BC065087.1 - 3 3278

View Full Product Details