Gene Transcripts

Gene Symbol
Slx1b
Entrez Gene ID
75764
Gene Name
SLX1 structure-specific endonuclease subunit homolog B (S. cerevisiae)
Gene Aliases
1110030E23Rik, 2410170E21Rik, 4833422P03Rik, AI853643, Giyd1, Giyd2
Chromosome Location
Chr. 7: 126688927 - 126695783 on Build GRCm38
UniGene ID
Mm.280573
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NR_033446.1 5 2867
GenBank mRNA AK003973.1 - 1 250
AK153779.1 - 1 849
AK163709.1 - 1 677


Non Gene Transcripts

Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID Transcript Location
GenBankmRNA AA097433.1 - 1 348 Chr. 7 - 126689525 - 126690047 [-]
GenBankmRNA BY704222.1 - 1 250 Chr. 7 - 126689537 - 126689949 [-]
GenBankmRNA AI595193.1 - 1 348 Chr. 7 - 126689526 - 126690047 [-]

View Full Product Details