Gene Transcripts
- Gene Symbol
- Snhg1
- Entrez Gene ID
- 83673
- Gene Name
- small nucleolar RNA host gene 1
- Gene Aliases
- Chromosome Location
- Chr. 19: 8723487 - 8726326 on Build GRCm38
- UniGene ID
- Mm.292489
- Species
- Mus musculus
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
GenBank mRNA |
AK036616.1 |
- |
1 |
1512 |
|
AK051045.1 |
- |
2 |
1298 |
|
Non Gene Transcripts
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
Transcript Location |
GenBankmRNA |
U40654.1 |
- |
1 |
1668 |
|
Chr. - - [-] |
View Full Product Details