Gene Transcripts

Gene Symbol
Shc2
Entrez Gene ID
216148
Gene Name
SHC (Src homology 2 domain containing) transforming protein 2
Gene Aliases
6720466E06, SCK, ShcB, Sli
Chromosome Location
Chr. 10: 79617934 - 79637918 on Build GRCm38
UniGene ID
Mm.39424
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001024539.1 NP_001019710.1 10 1250
GenBank mRNA AK032878.1 - 11 935

View Full Product Details