Gene Transcripts

Gene Symbol
Nipa1
Entrez Gene ID
233280
Gene Name
non imprinted in Prader-Willi/Angelman syndrome 1 homolog (human)
Gene Aliases
1110027G09Rik, A830014A18Rik, FSP3, Spg6
Chromosome Location
Chr. 7: 55978484 - 56019573 on Build GRCm38
UniGene ID
Mm.389901
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_153578.2 NP_705806.1 5 652
GenBank mRNA AK046619.1 - 5 667
AK166325.1 - 630
AK182759.1 - 3 400
AY098645.1 - 5 652
BC055828.1 - 5 652 4192279

View Full Product Details