Gene Transcripts

Gene Symbol
Slc26a9
Entrez Gene ID
320718
Gene Name
solute carrier family 26, member 9
Gene Aliases
E030002L01Rik
Chromosome Location
Chr. 1: 131744022 - 131771504 on Build GRCm38
UniGene ID
Mm.482704
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_177243.4 NP_796217.2 16, 17 1887
GenBank mRNA AK078966.1 - 16, 17 1888
AK086815.1 - 16, 17 1888
AY034145.1 - 15, 16 1800

View Full Product Details