Gene Transcripts

Gene Symbol
Tbc1d2
Entrez Gene ID
381605
Gene Name
TBC1 domain family, member 2
Gene Aliases
A630005A06Rik, BC022630, Gm1037, PARIS-1, PARIS1
Chromosome Location
Chr. 4: 46604390 - 46650877 on Build GRCm38
UniGene ID
Mm.331595
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_198664.3 NP_941066.3 9 2215
GenBank mRNA AK148254.1 - 9 2218
AK148289.1 - 9 2131
AK193678.1 - 1 43

View Full Product Details