Gene Transcripts

Gene Symbol
Shisa8
Entrez Gene ID
435145
Gene Name
shisa family member 8
Gene Aliases
Orf26
Chromosome Location
Chr. 15: 82206952 - 82212815 on Build GRCm38
UniGene ID
Mm.41289
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001207021.1 NP_001193950.1 2 1005
GenBank mRNA AW213062.1 - 2 171
BU558590.1 - 2 212

View Full Product Details