Gene Transcripts

Gene Symbol
Glipr1l1
Entrez Gene ID
69286
Gene Name
GLI pathogenesis-related 1 like 1
Gene Aliases
1700011E04Rik
Chromosome Location
Chr. 10: 112060189 - 112078510 on Build GRCm38
UniGene ID
Mm.47058
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_027018.1 NP_081294.1 1 270
GenBank mRNA AK005860.1 - 1 271

View Full Product Details