Gene Transcripts

Gene Symbol
RYK
Entrez Gene ID
6259
Gene Name
receptor-like tyrosine kinase
Gene Aliases
D3S3195, JTK5, JTK5A, RYK1
Chromosome Location
Chr. 3: 134157133 - 134250834 on Build GRCh38
UniGene ID
Hs.654562
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001005861.2 NP_001005861.1 8 1064
NM_002958.3 NP_002949.2 8 1055
XM_017007016.1 XP_016862505.1 3 561
GenBank mRNA BC021700.2 - 3 330
S59184.1 - 8 1088
X96588.1 - 9 960

View Full Product Details