Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_001007067.1 | NP_001007068.1 | 7 | 748 | |
NM_001007068.1 | NP_001007069.1 | 7 | 733 | ||
NM_001007069.1 | NP_001007070.1 | 7 | 748 | ||
NM_001007070.1 | NP_001007071.1 | 7 | 745 | ||
NM_005625.3 | NP_005616.2 | 7 | 751 | ||
XM_011517571.2 | XP_011515873.2 | 7 | 2120 | ||
XM_017013727.1 | XP_016869216.1 | 7 | 2123 | ||
XM_017013728.1 | XP_016869217.1 | 7 | 2120 | ||
GenBank mRNA | AF000652.1 | - | 7 | 749 | |
AF006636.1 | - | 7 | 676 | ||
AK128645.1 | - | 8 | 2175 | ||
AK295198.1 | - | 6 | 488 | ||
AK298697.1 | - | 7 | 563 | ||
AK300647.1 | - | 7 | 665 | ||
AK312274.1 | - | 7 | 659 | ||
BC013254.2 | - | 7 | 635 | ||
BC113674.1 | - | 7 | 695 | 8322726 | |
BC113676.1 | - | 7 | 695 | 8322728 | |
BC143915.1 | - | 7 | 692 | 9052433 | |
BC143916.1 | - | 7 | 695 | 9052434 | |
U83463.1 | - | 6 | 625 |