Gene Transcripts

Gene Symbol
Prss38
Entrez Gene ID
287358
Gene Name
protease, serine, 38
Gene Aliases
Mpn2
Chromosome Location
Chr. 10: 45723663 - 45726805 on Build Rnor_6.0
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq XM_006246538.2 XP_006246600.1 1 642
XM_220559.8 XP_220559.5 2 468

View Full Product Details