Gene Transcripts

Gene Symbol
TPMT
Entrez Gene ID
7172
Gene Name
thiopurine S-methyltransferase
Gene Aliases
TPMTD
Chromosome Location
Chr. 6: 18128311 - 18155165 on Build GRCh38
UniGene ID
Hs.444319
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq XM_011514839.2 XP_011513141.1 6 617
XM_017011241.1 XP_016866730.1 7 953
GenBank mRNA BC005339.1 - 6 569
BI598325.1 - 6 644

View Full Product Details