Gene Transcripts

Gene Symbol
Ttc29
Entrez Gene ID
291944
Gene Name
tetratricopeptide repeat domain 29
Gene Aliases
Chromosome Location
Chr. 19: 33206531 - 33424955 on Build Rnor_6.0
UniGene ID
Rn.215003
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001013890.1 NP_001013912.1 6 669
GenBank mRNA BC078968.1 - 6 669

View Full Product Details