Gene Transcripts

Gene Symbol
Olr97
Entrez Gene ID
293238
Gene Name
olfactory receptor 97
Gene Aliases
ratchr1-161209792-161208782_ORF
Chromosome Location
Chr. 1: 168500223 - 168501116 on Build Rnor_6.0
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq XM_006229923.1 XP_006229985.1 1 402

View Full Product Details