Gene Transcripts

Gene Symbol
SLC30A2
Entrez Gene ID
7780
Gene Name
solute carrier family 30 member 2
Gene Aliases
PP12488, TNZD, ZNT2, ZnT-2
Chromosome Location
Chr. 1: 26038022 - 26046138 on Build GRCh38
UniGene ID
Hs.143545
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001004434.2 NP_001004434.1 4 695
NM_032513.4 NP_115902.1 3 548
GenBank mRNA AF370409.1 - 4 542
AK094027.1 - 4 511
BC006251.2 - 3 523 3948165
DR004258.1 - 4 702

View Full Product Details