Gene Transcripts

Gene Symbol
Lpar4
Entrez Gene ID
302378
Gene Name
lysophosphatidic acid receptor 4
Gene Aliases
Gpr23, RGD1563686
Chromosome Location
Chr. X: 78042859 - 78056347 on Build Rnor_6.0
UniGene ID
Rn.105026
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001106940.1 NP_001100410.1 3 1259

View Full Product Details