Gene Transcripts

Gene Symbol
Zc3h7b
Entrez Gene ID
315158
Gene Name
zinc finger CCCH-type containing 7B
Gene Aliases
RGD1307064
Chromosome Location
Chr. 7: 122978987 - 123027166 on Build Rnor_6.0
UniGene ID
Rn.91605
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001130695.1 NP_001124167.1 20 2440
XM_006242085.2 XP_006242147.1 21 2724
XM_008765719.1 XP_008763941.1 21 3810

View Full Product Details