Gene Transcripts

Gene Symbol
Mcat
Entrez Gene ID
315173
Gene Name
malonyl CoA:ACP acyltransferase (mitochondrial)
Gene Aliases
RGD1306698
Chromosome Location
Chr. 7: 124412762 - 124424966 on Build Rnor_6.0
UniGene ID
Rn.23115
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001191788.1 NP_001178717.1 4 802
XM_006242091.1 XP_006242153.1 5 1063
GenBank mRNA FQ232982.1 - 4 801

View Full Product Details