Gene Transcripts

Gene Symbol
Spetex-2H
Entrez Gene ID
361008
Gene Name
Spetex-2H protein
Gene Aliases
Spergen-2H
Chromosome Location
Chr. : - on Build
UniGene ID
Rn.162039
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001009969.1 NP_001009969.1 1, 2 94
GenBank mRNA AB180083.1 - 1, 2 94

Gene Symbol
Spetex-2B
Entrez Gene ID
497084
Gene Name
Spetex-2B protein
Gene Aliases
Chromosome Location
Chr. : - on Build
UniGene ID
Rn.198259
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001011697.1 NP_001011697.1 1, 2 94
GenBank mRNA AB180077.1 - 1, 2 94

Gene Symbol
Spetex-2D
Entrez Gene ID
364260
Gene Name
Spetex-2D protein
Gene Aliases
Chromosome Location
Chr. : - on Build
UniGene ID
Rn.198259
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001011701.1 NP_001011701.1 1, 2 94
GenBank mRNA AB180079.1 - 1, 2 94

View Full Product Details