Gene Transcripts

Gene Symbol
Spetex-2H
Entrez Gene ID
361008
Gene Name
Spetex-2H protein
Gene Aliases
Spergen-2H
Chromosome Location
Chr. : - on Build
UniGene ID
Rn.162039
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001009969.1 NP_001009969.1 4 499
GenBank mRNA AB180083.1 - 4 499

View Full Product Details