Gene Transcripts

Gene Symbol
Siva1
Entrez Gene ID
362791
Gene Name
SIVA1, apoptosis-inducing factor
Gene Aliases
Siva
Chromosome Location
Chr. 6: 137210007 - 137214379 on Build Rnor_6.0
UniGene ID
Rn.23843
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001100982.1 NP_001094452.1 2 274
XM_006240645.2 XP_006240707.1 2 388
GenBank mRNA BC088866.1 - 2 274

View Full Product Details