Gene Transcripts

Gene Symbol
Lsm7
Entrez Gene ID
362829
Gene Name
LSM7 homolog, U6 small nuclear RNA associated (S. cerevisiae)
Gene Aliases
Chromosome Location
Chr. 7: 11724962 - 11727388 on Build Rnor_6.0
UniGene ID
Rn.25010
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001108732.1 NP_001102202.1 2 98
XM_006240958.2 XP_006241020.1 2 110
XM_008765118.1 XP_008763340.1 2 100

View Full Product Details