Gene Transcripts

Gene Symbol
BTAF1
Entrez Gene ID
9044
Gene Name
B-TFIID TATA-box binding protein associated factor 1
Gene Aliases
MOT1, TAF(II)170, TAF172, TAFII170
Chromosome Location
Chr. 10: 91923769 - 92030998 on Build GRCh38
UniGene ID
Hs.500526
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_003972.2 NP_003963.1 14 1720
XM_011540326.1 XP_011538628.1 14 1819
XM_011540327.2 XP_011538629.1 11 1485
XM_011540328.2 XP_011538630.1 11 1313
XM_011540329.2 XP_011538631.1 14 1930
XM_017016877.1 XP_016872366.1 13 1782
XM_017016878.1 XP_016872367.1 13 1612
XM_017016879.1 XP_016872368.1 13 1736
XM_017016880.1 XP_016872369.1 13 1740
XM_017016881.1 XP_016872370.1 12 1589
GenBank mRNA AF038362.1 - 14 1739
AJ001017.2 - 14 1634
BC030273.1 - 8 967
BC047009.2 - 2 212
BC112201.1 - 14 1704 8327669

View Full Product Details