Gene Transcripts

Gene Symbol
BTAF1
Entrez Gene ID
9044
Gene Name
B-TFIID TATA-box binding protein associated factor 1
Gene Aliases
MOT1, TAF(II)170, TAF172, TAFII170
Chromosome Location
Chr. 10: 91923769 - 92030998 on Build GRCh38
UniGene ID
Hs.500526
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_003972.2 NP_003963.1 37 5357
XM_011540326.1 XP_011538628.1 37 5456
XM_011540327.2 XP_011538629.1 34 5122
XM_011540328.2 XP_011538630.1 34 4950
XM_017016877.1 XP_016872366.1 36 5419
XM_017016878.1 XP_016872367.1 36 5249
XM_017016879.1 XP_016872368.1 36 5373
XM_017016880.1 XP_016872369.1 36 5377
XM_017016881.1 XP_016872370.1 35 5226
GenBank mRNA AB208843.1 - 25 3751
AF038362.1 - 37 5376
AJ001017.2 - 37 5271
AK303554.1 - 15 2034
BC029930.1 - 13 1752
BC112201.1 - 37 5341 8327669

View Full Product Details