Gene Transcripts

Gene Symbol
Ube4b
Entrez Gene ID
298652
Gene Name
ubiquitination factor E4B
Gene Aliases
ufd2a
Chromosome Location
Chr. 5: 166156033 - 166259069 on Build Rnor_6.0
UniGene ID
Rn.19289
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001271198.1 NP_001258127.1 25 4051
XM_006239387.2 XP_006239449.1 27 4585

View Full Product Details