Gene Transcripts

Gene Symbol
Ly49s7
Entrez Gene ID
494203
Gene Name
Ly49 stimulatory receptor 7
Gene Aliases
Chromosome Location
Chr. : - on Build
UniGene ID
Rn.129029
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001009494.1 NP_001009494.1 4 375
GenBank mRNA AY649833.1 - 4 375
AY649840.1 - 4 367

View Full Product Details