Gene Transcripts

Gene Symbol
Prss33
Entrez Gene ID
497873
Gene Name
protease, serine, 33
Gene Aliases
Chromosome Location
Chr. 10: 13173509 - 13176288 on Build Rnor_6.0
UniGene ID
Rn.218707
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001109027.1 NP_001102497.1 4 304
XM_006245874.2 XP_006245936.1 5 594

View Full Product Details