Gene Transcripts

Gene Symbol
Pex10
Entrez Gene ID
680424
Gene Name
peroxisomal biogenesis factor 10
Gene Aliases
Chromosome Location
Chr. 5: 172469978 - 172475144 on Build Rnor_6.0
UniGene ID
Rn.111
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001109405.1 NP_001102875.1 2 227

View Full Product Details