Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_001134797.1 | NP_001128269.1 | 5 | 501 | |
XM_006252813.2 | XP_006252875.1 | 3 | 482 | ||
XM_006252814.2 | XP_006252876.1 | 4 | 393 | ||
XM_006252816.2 | XP_006252878.1 | 3 | 322 | ||
XM_008771149.1 | XP_008769371.1 | 3 | 345 |