Gene Transcripts

Gene Symbol
Pramef17
Entrez Gene ID
691139
Gene Name
PRAME family member 17
Gene Aliases
Chromosome Location
Chr. 5: 162440137 - 162443016 on Build Rnor_6.0
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq XM_001076970.3 XP_001076970.1 3 995

View Full Product Details