Gene Transcripts

Gene Symbol
XAF1
Entrez Gene ID
679600
Gene Name
XIAP associated factor-1
Gene Aliases
Chromosome Location
Chr. 10: 58860647 - 58872782 on Build Rnor_6.0
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq XM_006246855.2 XP_006246917.1 4, 5 736

View Full Product Details