Gene Transcripts

Gene Symbol
SHISA8
Entrez Gene ID
440829
Gene Name
shisa family member 8
Gene Aliases
C22orf17, Orf26
Chromosome Location
Chr. 22: 41907180 - 41915093 on Build GRCh38
UniGene ID
Hs.526704
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001207020.1 NP_001193949.1 4 831
XM_006724254.3 XP_006724317.1 4 1542
XM_006724255.2 XP_006724318.1 4 1434
XM_006724256.3 XP_006724319.1 4 1079

View Full Product Details