Gene Transcripts
- Gene Symbol
- Itln1
- Entrez Gene ID
- 16429
- Gene Name
- intelectin 1 (galactofuranose binding)
- Gene Aliases
- IntL, Itln, Itln2, Itln5, Itlna, Lfr
- Chromosome Location
- Chr. 1: 171518123 - 171535325 on Build GRCm38
- UniGene ID
- Mm.247426
- Species
- Mus musculus
- Gene Symbol
- Itlnb
- Entrez Gene ID
- 493583
- Gene Name
- intelectin b
- Gene Aliases
- Itln1b, Itln2
- Chromosome Location
- Chr. : - on Build
- UniGene ID
- Mm.247426
- Species
- Mus musculus
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
GenBank mRNA |
BC145376.1 |
- |
2, 3 |
84 |
|
View Full Product Details