Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_001168388.2 | NP_001161860.1 | 2 | 789 | |
NM_001168389.2 | NP_001161861.2 | 2 | 772 | ||
NM_006079.4 | NP_006070.2 | 2 | 928 | ||
GenBank mRNA | AF109161.1 | - | 2 | 898 | |
BC004377.1 | - | 2 | 881 | 3640855 | |
U65093.1 | - | 3 | 710 |