Gene Transcripts

Gene Symbol
Tmem97
Entrez Gene ID
303330
Gene Name
transmembrane protein 97
Gene Aliases
RGD1307423
Chromosome Location
Chr. 10: 65811455 - 65820538 on Build Rnor_6.0
UniGene ID
Rn.20057
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001008334.1 NP_001008335.1 1 98
GenBank mRNA BC085344.1 - 1 98
FQ214423.1 - 1 140

View Full Product Details