Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_000542.3 | NP_000533.3 | 4 | 370 | |
NM_198843.2 | NP_942140.2 | 4 | 370 | ||
XM_005264487.2 | XP_005264544.1 | 3 | 285 | ||
XM_005264488.3 | XP_005264545.2 | 3 | 250 | ||
XM_005264490.4 | XP_005264547.2 | 3 | 250 | ||
XM_017004702.1 | XP_016860191.1 | 3 | 285 | ||
XM_017004703.1 | XP_016860192.1 | 3 | 250 | ||
GenBank mRNA | AK314947.1 | - | 3 | 250 | |
AL548140.3 | - | 4 | 370 | ||
BC032785.1 | - | 3 | 237 | 5179408 | |
BI821350.1 | - | 240 | |||
DQ317589.1 | - | 3 | 223 | ||
J02761.1 | - | 3 | 247 |