Gene Transcripts

Gene Symbol
Dhx9
Entrez Gene ID
13211
Gene Name
DEAH (Asp-Glu-Ala-His) box polypeptide 9
Gene Aliases
AI326842, Ddx9, HEL-5, NDHII, RHA, mHEL-5
Chromosome Location
Chr. 1: 153455756 - 153487686 on Build GRCm38
UniGene ID
Mm.20000
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_007842.2 NP_031868.2 5 624
GenBank mRNA AK013423.1 - 5 628
AK141824.1 - 5 619
AK148423.1 - 5 620
BC033911.1 - 5 627
BC089159.1 - 5 618
U91922.1 - 5 581

View Full Product Details