Gene Transcripts

Gene Symbol
Tmprss9
Entrez Gene ID
314636
Gene Name
transmembrane protease, serine 9
Gene Aliases
Chromosome Location
Chr. 7: 11678591 - 11708980 on Build Rnor_6.0
UniGene ID
Rn.123548
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001047100.2 NP_001040565.2 8 1063
XM_006240931.1 XP_006240993.1 10 1667
GenBank mRNA AB109392.1 - 9 1177

View Full Product Details