Gene Transcripts

Gene Symbol
CNPPD1
Entrez Gene ID
27013
Gene Name
cyclin Pas1/PHO80 domain containing 1
Gene Aliases
C2orf24, CGI-57
Chromosome Location
Chr. 2: 219171897 - 219178174 on Build GRCh38
UniGene ID
Hs.4973
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001321389.1 NP_001308318.1 4 621
NM_001321390.1 NP_001308319.1 4 266
NM_001321391.1 NP_001308320.1 4 300
NM_015680.5 NP_056495.3 3 358
NR_135628.1 4 300
NR_135629.1 3 358
GenBank mRNA AF151815.1 - 4 525
AK075274.1 - 3 314
AK311656.1 - 2 845
AK314975.1 - 3 301
AY007160.1 - 3 331
BC001393.2 - 3 281 3051397
BC012821.2 - 3 330 2958621
BI912613.1 - 4 267
CR457401.1 - 3 193
CX166359.1 - 4 617
DB477110.1 - 3 345
HY106884.1 - 3 358

View Full Product Details