Gene Transcripts
- Gene Symbol
- YPEL5
- Entrez Gene ID
- 51646
- Gene Name
- yippee like 5
- Gene Aliases
- CGI-127
- Chromosome Location
- Chr. 2: 30146568 - 30160533 on Build GRCh38
- UniGene ID
- Hs.515890
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NM_001127399.1 |
NP_001120871.1
|
3, 4 |
409 |
|
NM_001127400.1 |
NP_001120872.1
|
3, 4 |
645 |
|
NM_001127401.1 |
NP_001120873.1
|
4, 5 |
706 |
|
NM_016061.2 |
NP_057145.1
|
2, 3 |
348 |
|
XM_017004318.1 |
XP_016859807.1
|
4, 5 |
944 |
|
XM_017004319.1 |
XP_016859808.1
|
3, 4 |
382 |
|
XM_017004320.1 |
XP_016859809.1
|
3, 4 |
883 |
|
XM_017004321.1 |
XP_016859810.1
|
3, 4 |
575 |
|
XM_017004322.1 |
XP_016859811.1
|
3, 4 |
806 |
|
GenBank mRNA |
AB098739.1 |
- |
1, 2 |
145 |
|
AF135161.1 |
- |
2, 3 |
252 |
|
AF151885.1 |
- |
2, 3 |
270 |
|
AF172940.1 |
- |
2, 3 |
272 |
|
AK025730.1 |
- |
3, 4 |
286 |
|
BC000836.1 |
- |
3, 4 |
223 |
3456031 |
BC047237.1 |
- |
3, 4 |
806 |
5269563 |
View Full Product Details