Gene Transcripts

Gene Symbol
PCDHB2
Entrez Gene ID
56133
Gene Name
protocadherin beta 2
Gene Aliases
PCDH-BETA2
Chromosome Location
Chr. 5: 141094607 - 141097380 on Build GRCh38
UniGene ID
Hs.533023
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_018936.3 NP_061759.1 1 1233
GenBank mRNA AB209745.1 - 1 1009
AF152495.1 - 1 1049
AF217756.1 - 1 1049
AK027526.1 - 1 1204
AK314642.1 - 1 1187
BC098347.1 - 1 1096 40007951
BC098575.1 - 1 1197 30921306

View Full Product Details