Gene Transcripts

Gene Symbol
MYCT1
Entrez Gene ID
80177
Gene Name
myc target 1
Gene Aliases
MTLC
Chromosome Location
Chr. 6: 152697895 - 152747171 on Build GRCh38
UniGene ID
Hs.18160
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_025107.2 NP_079383.2 2 345
XM_011536149.2 XP_011534451.1 2 345
XM_011536150.2 XP_011534452.1 2 345
XM_011536151.2 XP_011534453.1 2 345
XM_011536152.2 XP_011534454.1 2 345
XM_017011319.1 XP_016866808.1 2 345
GenBank mRNA AF527367.1 - 2 339
AK024922.1 - 2 337
AL834500.1 - 2 244
BC022556.1 - 2 294
BC128463.1 - 2 337 40126671
BC128464.1 - 2 337 40126673
DA863771.1 - 2 345
GU997693.1 - 2 333

View Full Product Details