Gene Transcripts

Gene Symbol
TTC29
Entrez Gene ID
83894
Gene Name
tetratricopeptide repeat domain 29
Gene Aliases
NYD-SP14, TBPP2A
Chromosome Location
Chr. 4: 146706638 - 146945886 on Build GRCh38
UniGene ID
Hs.378893
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001300761.2 NP_001287690.1 8 1039
NM_001317806.1 NP_001304735.1 7 938
NM_031956.3 NP_114162.2 7 938
NR_133922.1 7 938
XM_005263270.1 XP_005263327.1 8 1021
XM_006714335.1 XP_006714398.1 8 1021
XM_006714336.1 XP_006714399.1 8 1021
XM_006714339.2 XP_006714402.1 7 835
XM_011532310.2 XP_011530612.1 8 1021
GenBank mRNA AF345910.1 - 7 889
AK225725.1 - 7 885
AK314066.1 - 7 934
BC024193.2 - 7 880 4825866
EF432564.1 - 5 707

View Full Product Details