Gene Transcripts

Gene Symbol
PAGE5
Entrez Gene ID
90737
Gene Name
PAGE family member 5
Gene Aliases
CT16, CT16.1, CT16.2, GAGEE1, PAGE-5
Chromosome Location
Chr. X: 55220347 - 55224108 on Build GRCh38
UniGene ID
Hs.668491
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001013435.2 NP_001013453.1 4 356
NM_130467.4 NP_569734.2 4 555
GenBank mRNA AJ344352.1 - 4 525
BC009230.2 - 4 544 3955765
BC110803.1 - 4 290 3922538
BQ437151.1 - 4 344

Gene Symbol
LOC100421746
Entrez Gene ID
100421746
Gene Name
PAGE family member 5 pseudogene
Gene Aliases
Chromosome Location
Chr. X: 55280088 - 55288786 on Build GRCh38
UniGene ID
Hs.662612
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NR_130179.1 4 318

View Full Product Details