Gene Transcripts
- Gene Symbol
- SPAG17
- Entrez Gene ID
- 200162
- Gene Name
- sperm associated antigen 17
- Gene Aliases
- CT143, PF6
- Chromosome Location
- Chr. 1: 117953665 - 118185225 on Build GRCh38
- UniGene ID
- Hs.528821
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NM_206996.2 |
NP_996879.1
|
21, 22 |
3114 |
|
XM_006710426.3 |
XP_006710489.1
|
21, 22 |
3114 |
|
XM_006710427.3 |
XP_006710490.1
|
20, 21 |
2919 |
|
XM_006710428.3 |
XP_006710491.1
|
8, 9 |
1185 |
|
XM_006710429.3 |
XP_006710492.1
|
5, 6 |
797 |
|
XM_011540934.1 |
XP_011539236.1
|
21, 22 |
3114 |
|
XM_011540935.2 |
XP_011539237.1
|
20, 21 |
2953 |
|
XM_011540936.2 |
XP_011539238.1
|
12, 13 |
1772 |
|
XM_011540937.2 |
XP_011539239.1
|
12, 13 |
1938 |
|
XM_011540939.2 |
XP_011539241.1
|
9, 10 |
1223 |
|
XM_011540941.2 |
XP_011539243.1
|
8, 9 |
1233 |
|
XM_011540942.2 |
XP_011539244.1
|
8, 9 |
1041 |
|
GenBank mRNA |
AL833485.1 |
- |
7, 8 |
884 |
|
AY555274.1 |
- |
21, 22 |
3100 |
|
BX648110.1 |
- |
21, 22 |
3115 |
|
View Full Product Details