Gene Transcripts
- Gene Symbol
- TAS2R45
- Entrez Gene ID
- 259291
- Gene Name
- taste 2 receptor member 45
- Gene Aliases
- GPR59, T2R45, ZG24P
- Chromosome Location
- Chr. : - on Build
- UniGene ID
- Hs.287378
- Species
- Homo sapiens
- Gene Symbol
- TAS2R43
- Entrez Gene ID
- 259289
- Gene Name
- taste 2 receptor member 43
- Gene Aliases
- T2R43, T2R52
- Chromosome Location
- Chr. 12: 11091287 - 11092313 on Build GRCh38
- UniGene ID
- Hs.688195
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
GenBank mRNA |
BC101155.1 |
- |
3 |
574 |
|
View Full Product Details